Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt72
Entrez Gene ID
406227
Gene Name
keratin 72, type II
Gene Aliases
Kb35
Chromosome Location
Chr.7: 143382606 - 143392777 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008809.1 NP_001008809.1 7 - 8 1336 141

View Full Product Details