Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc8d
Entrez Gene ID
305131
Gene Name
leucine rich repeat containing 8 family, member D
Gene Aliases
Lrrc5
Chromosome Location
Chr.14: 5085606 - 5166691 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008338.1 NP_001008339.2 2 - 2 2941 75
XM_006250556.2 XP_006250618.1 2 - 2 2948 75
GenBank mRNA BC085783.1 - 2 - 2 2941 75

View Full Product Details