The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013065.1 | NP_001013083.1 | 1 - 2 | 2234 | 80 |
XM_006228718.2 | XP_006228780.1 | 2 - 3 | 2829 | 80 | |
XM_006228719.2 | XP_006228781.1 | 2 - 3 | 2663 | 80 | |
XM_008759123.1 | XP_008757345.1 | 2 - 3 | 2989 | 80 | |
GenBank mRNA | BC081922.1 | - | 1 - 2 | 2234 | 80 |
BC100147.1 | - | 2 - 3 | 1978 | 80 |