Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fos
Entrez Gene ID
314322
Gene Name
FBJ osteosarcoma oncogene
Gene Aliases
c-fos
Chromosome Location
Chr.6: 109300433 - 109303299 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022197.2 NP_071533.1 3 - 4 658 82
GenBank mRNA FQ228884.1 - 2 - 3 311 82
X06769.1 - - 640 82

View Full Product Details