Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt72
Entrez Gene ID
406227
Gene Name
keratin 72, type II
Gene Aliases
Kb35
Chromosome Location
Chr.7: 143382606 - 143392777 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008809.1 NP_001008809.1 9 - 9 1625 66

View Full Product Details