Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RT1-CE15
Entrez Gene ID
414789
Gene Name
RT1 class I, locus CE15
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008838.1 NP_001008838.1 1 - 1 277 65

Gene Symbol
LOC103694386
Entrez Gene ID
103694386
Gene Name
RT1 class I histocompatibility antigen, AA alpha chain
Gene Aliases
-
Chromosome Location
Chr.20: 2706819 - 2713704 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772782.1 XP_008771004.1 2 - 2 442 65

View Full Product Details