The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024886.1 | NP_001020057.1 | 2 - 3 | 742 | 67 |
XM_008759750.1 | XP_008757972.1 | 2 - 3 | 668 | 67 | |
GenBank mRNA | BC097462.1 | - | 2 - 3 | 742 | 67 |