The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013034.1 | NP_001013052.1 | 7 - 8 | 824 | 67 |
GenBank mRNA | BC079292.1 | - | 7 - 8 | 824 | 67 |