Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shisa4
Entrez Gene ID
360848
Gene Name
shisa family member 4
Gene Aliases
Prmp, RGD1312041, Tmem58
Chromosome Location
Chr.13: 52193989 - 52197205 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077826.1 NP_001071294.1 3 - 4 379 110
GenBank mRNA BC158557.1 - - 482 110
EF044301.1 - 3 - 4 379 110
FQ213412.1 - 3 - 4 390 110

View Full Product Details