The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106640.1 | NP_001100110.1 | 2 - 2 | 1614 | 126 |
XM_006237949.2 | XP_006238011.1 | 1 - 1 | 1351 | 126 | |
XM_008763592.1 | XP_008761814.1 | 2 - 2 | 1865 | 126 | |
XM_008763593.1 | XP_008761815.1 | 2 - 2 | 1367 | 126 |