Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vom2r30
Entrez Gene ID
289295
Gene Name
vomeronasal 2 receptor, 30
Gene Aliases
RGD1561133
Chromosome Location
Chr.1: 74770009 - 74791415 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099467.1 NP_001092937.1 3 - 4 1295 136

View Full Product Details