Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nxnl1
Entrez Gene ID
306342
Gene Name
nucleoredoxin-like 1
Gene Aliases
Txnl6
Chromosome Location
Chr.16: 19987113 - 19997201 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271341.1 NP_001258270.1 1 - 1 136 95
XM_006252893.2 XP_006252955.1 1 - 1 621 95
XM_006252894.2 XP_006252956.1 1 - 1 621 95
XM_008771104.1 XP_008769326.1 1 - 1 621 95
GenBank mRNA EU861029.1 - 1 - 1 148 95

View Full Product Details