Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Uchl3
Entrez Gene ID
498560
Gene Name
ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase)
Gene Aliases
RGD1561196
Chromosome Location
Chr.15: 86153628 - 86198127 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110165.1 NP_001103635.1 8 - 9 686 104
GenBank mRNA AB043959.1 - 8 - 9 611 104

View Full Product Details