Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Cers3
Entrez Gene ID
499174
Gene Name
ceramide synthase 3
Gene Aliases
Lass3, RGD1564356
Chromosome Location
Chr.1: 127706618 - 127781041 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127561.1 NP_001121033.1 - 541 146
GenBank mRNA BC160913.1 - - 541 146

View Full Product Details