Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agmo
Entrez Gene ID
362732
Gene Name
alkylglycerol monooxygenase
Gene Aliases
RGD1312038, Tmem195
Chromosome Location
Chr.6: 56846859 - 57193918 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135899.1 NP_001129371.1 5 - 6 742 110
GenBank mRNA BC127545.1 - 5 - 6 742 110
FQ211046.1 - 5 - 6 798 110
FQ219104.1 - 5 - 6 786 110

View Full Product Details