Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nkx6-2
Entrez Gene ID
309095
Gene Name
NK6 homeobox 2
Gene Aliases
-
Chromosome Location
Chr.1: 211922389 - 211923929 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107558.2 NP_001101028.1 1 - 2 642 89

View Full Product Details