The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012940.2 | NP_037072.1 | 1 - 2 | 373 | 69 |
GenBank mRNA | U09540.1 | - | - | 373 | 69 |