Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gdi2
Entrez Gene ID
29662
Gene Name
GDP dissociation inhibitor 2
Gene Aliases
Gdi3
Chromosome Location
Chr.17: 70299177 - 70325864 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017276.2 NP_058972.2 1 - 2 143 94
GenBank mRNA BC061767.1 - 1 - 2 143 94
X74401.1 - 1 - 2 73 94

View Full Product Details