Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nppb
Entrez Gene ID
25105
Gene Name
natriuretic peptide B
Gene Aliases
BNP, Bnf
Chromosome Location
Chr.5: 164796176 - 164797538 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031545.1 NP_113733.1 2 - 3 402 93
XM_006239376.1 XP_006239438.1 2 - 3 93
GenBank mRNA FQ228997.1 - - 93
M25297.1 - 2 - 3 402 93

View Full Product Details