Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim58
Entrez Gene ID
303167
Gene Name
tripartite motif-containing 58
Gene Aliases
RGD1566174
Chromosome Location
Chr.10: 44271587 - 44282726 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191641.1 NP_001178570.1 5 - 6 869 60

View Full Product Details