Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ict1
Entrez Gene ID
303673
Gene Name
immature colon carcinoma transcript 1
Gene Aliases
-
Chromosome Location
Chr.10: 103956019 - 103962515 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191656.1 NP_001178585.1 1 - 2 190 76

View Full Product Details