The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195807.1 | NP_001182736.1 | 2 - 3 | 426 | 77 |
XM_006239955.2 | XP_006240017.1 | 2 - 3 | 797 | 77 | |
XM_006239956.2 | XP_006240018.1 | 2 - 3 | 796 | 77 | |
XM_006239957.2 | XP_006240019.1 | 2 - 3 | 796 | 77 | |
XM_006239958.2 | XP_006240020.1 | 2 - 3 | 796 | 77 | |
GenBank mRNA | CK596582.1 | - | 2 - 3 | 426 | 77 |