Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sohlh1
Entrez Gene ID
362085
Gene Name
spermatogenesis and oogenesis specific basic helix-loop-helix 1
Gene Aliases
RGD1564440
Chromosome Location
Chr.3: 3291084 - 3295101 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191852.1 NP_001178781.1 3 - 4 353 82

View Full Product Details