Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Xbp1
Entrez Gene ID
289754
Gene Name
X-box binding protein 1
Gene Aliases
HTF
Chromosome Location
Chr.14: 85753736 - 85758820 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271731.1 NP_001258660.1 5 - 5 522 68
GenBank mRNA CB691171.1 - 3 - 3 120 68

View Full Product Details