Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX22
Entrez Gene ID
851244
Gene Name
Gene Aliases
YAF5, YAL055W
Chromosome Location
Chr.I: 42177 - 42719 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178198.1 NP_009346.1 1 - 1 447 82

View Full Product Details