Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSY4
Entrez Gene ID
852234
Gene Name
Gene Aliases
HSM6, YBL046W
Chromosome Location
Chr.II: 132424 - 133749 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178286.1 NP_009507.1 1 - 1 1251 92

View Full Product Details