Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TCM62
Entrez Gene ID
852332
Gene Name
Gene Aliases
YBR044C
Chromosome Location
Chr.II: 324341 - 326059 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178392.2 NP_009600.2 1 - 1 1548 75

View Full Product Details