Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MUM2
Entrez Gene ID
852348
Gene Name
Gene Aliases
YBR057C
Chromosome Location
Chr.II: 352193 - 353293 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178405.1 NP_009613.1 1 - 1 1049 92

View Full Product Details