Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECM33
Entrez Gene ID
852370
Gene Name
Gene Aliases
YBR078W
Chromosome Location
Chr.II: 393123 - 394742 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178426.2 NP_009634.2 1 - 2 52 102

View Full Product Details