Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD16
Entrez Gene ID
852411
Gene Name
Gene Aliases
PSO5, YBR114W
Chromosome Location
Chr.II: 467248 - 469620 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178462.1 NP_009672.1 1 - 1 2155 87

View Full Product Details