Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SWD3
Entrez Gene ID
852472
Gene Name
Gene Aliases
CPS30, SAF35, YBR175W
Chromosome Location
Chr.II: 582408 - 583355 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178523.1 NP_009734.1 1 - 1 852 99

View Full Product Details