Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRT2
Entrez Gene ID
852548
Gene Name
Gene Aliases
DPH7, ERE1, YBR246W
Chromosome Location
Chr.II: 711591 - 712754 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178594.1 NP_009805.1 1 - 1 1052 92

View Full Product Details