Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BIT2
Entrez Gene ID
852573
Gene Name
Gene Aliases
YBR270C
Chromosome Location
Chr.II: 742761 - 744398 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178618.2 NP_009829.2 1 - 1 1448 100

View Full Product Details