Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SOL2
Entrez Gene ID
850437
Gene Name
Gene Aliases
YCR073W-A, YCRX13W
Chromosome Location
Chr.III: 246963 - 247910 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178815.1 NP_009999.2 1 - 1 749 86

View Full Product Details