Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRX6
Entrez Gene ID
851551
Gene Name
Gene Aliases
YDL010W
Chromosome Location
Chr.IV: 432330 - 433025 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180069.1 NP_010274.1 1 - 1 250 71

View Full Product Details