Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDE2
Entrez Gene ID
851474
Gene Name
Gene Aliases
NDH2, YDL085W
Chromosome Location
Chr.IV: 303211 - 304848 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180144.1 NP_010198.1 1 - 1 1550 88

View Full Product Details