Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENT1
Entrez Gene ID
851392
Gene Name
Gene Aliases
YDL161W
Chromosome Location
Chr.IV: 167714 - 169078 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180221.1 NP_010120.1 1 - 1 1047 95

View Full Product Details