Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRS1
Entrez Gene ID
851607
Gene Name
Gene Aliases
GCD5, YDR037W
Chromosome Location
Chr.IV: 525440 - 527215 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180345.1 NP_010322.1 1 - 1 1650 71

View Full Product Details