Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMN2
Entrez Gene ID
851685
Gene Name
Gene Aliases
YDR107C
Chromosome Location
Chr.IV: 669016 - 671034 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180415.1 NP_010392.1 1 - 1 1553 97

View Full Product Details