Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RUB1
Entrez Gene ID
851717
Gene Name
Gene Aliases
YDR139C
Chromosome Location
Chr.IV: 733618 - 733924 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180446.3 NP_010423.4 1 - 2 146 87

View Full Product Details