Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YRA1
Entrez Gene ID
851988
Gene Name
Gene Aliases
SHE11, YDR381W
Chromosome Location
Chr.IV: 1236558 - 1238004 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180689.1 NP_010669.1 1 - 2 285 66

View Full Product Details