Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPP2B
Entrez Gene ID
851990
Gene Name
Gene Aliases
RPL45, YDR382W, YPA1
Chromosome Location
Chr.IV: 1239492 - 1239824 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180690.3 NP_010670.3 1 - 1 148 85

View Full Product Details