Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DFM1
Entrez Gene ID
852020
Gene Name
Gene Aliases
YDR411C
Chromosome Location
Chr.IV: 1293369 - 1294394 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180719.1 NP_010699.1 1 - 1 952 74

View Full Product Details