Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYF1
Entrez Gene ID
852025
Gene Name
Gene Aliases
NTC90, YDR416W
Chromosome Location
Chr.IV: 1298432 - 1301011 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180724.1 NP_010704.1 1 - 1 2444 86

View Full Product Details