Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX41
Entrez Gene ID
852035
Gene Name
Gene Aliases
YDR425W
Chromosome Location
Chr.IV: 1320064 - 1321941 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180733.1 NP_010713.1 1 - 1 1554 75

View Full Product Details