Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MZM1
Entrez Gene ID
852104
Gene Name
Gene Aliases
AIM8, FMP36, YDR493W
Chromosome Location
Chr.IV: 1436217 - 1436588 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180801.1 NP_010781.1 1 - 1 149 96

View Full Product Details