Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRT13
Entrez Gene ID
856796
Gene Name
Gene Aliases
YER066W
Chromosome Location
Chr.V: 290242 - 290799 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178957.1 NP_010988.1 1 - 1 455 85

View Full Product Details