Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAB1
Entrez Gene ID
856912
Gene Name
Gene Aliases
YER165W
Chromosome Location
Chr.V: 510373 - 512106 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179055.1 NP_011092.1 1 - 1 1650 80

View Full Product Details