Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LPD1
Entrez Gene ID
850527
Gene Name
Gene Aliases
HPD1, YFL018C
Chromosome Location
Chr.VI: 101628 - 103127 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179948.1 NP_116635.1 1 - 1 1446 67

View Full Product Details