Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EMP47
Entrez Gene ID
850496
Gene Name
Gene Aliases
YFL048C
Chromosome Location
Chr.VI: 38843 - 40180 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179919.1 NP_116606.1 1 - 1 1148 110

View Full Product Details